Learn More
MSC E.R. Wagner Swivel Top Plate Caster, Thermoplastic Rubber, 3-1/2 in Wheel
Catalog No. 5000511751
Encompass
This swivel top plate caster features a thermoplastic rubber wheel and a ball bearing. It is designed for various applications, including food service and general industrial/commercial environments, offering a load capacity of 155 lb.
  • Thermoplastic rubber wheel material
  • 3-1/2 in wheel diameter
  • 1-1/4 in wheel width
  • 155 lb load capacity
  • Ball bearing type
  • 3-1/8 x 4-1/8 in plate size
  • 4-5/8 in overall height
  • Zinc finish/coating
  • Suitable for food service and general industrial/commercial applications
  • Top plate mount style
  • Swivel style
Catalog No. 50-005-11751 Supplier MSC Supplier No. 57077695
This swivel top plate caster features a thermoplastic rubber wheel and a ball bearing. It is designed for various applications, including food service and general industrial/commercial environments, offering a load capacity of 155 lb.
  • Thermoplastic rubber wheel material
  • 3-1/2 in wheel diameter
  • 1-1/4 in wheel width
  • 155 lb load capacity
  • Ball bearing type
  • 3-1/8 x 4-1/8 in plate size
  • 4-5/8 in overall height
  • Zinc finish/coating
  • Suitable for food service and general industrial/commercial applications
  • Top plate mount style
  • Swivel style