Learn More
MSC E.R. Wagner Swivel Top Plate Caster, Thermoplastic Rubber, 6 in Wheel, 280 lb Capacity, Nickel
Catalog No. 5000513261
Encompass
This swivel top plate caster features a thermoplastic rubber wheel with a 6 in diameter and 1-1/2 in width. It offers a load capacity of 280 lb and an overall height of 7-3/8 in, making it suitable for various applications on hard surfaces. The caster is constructed with hardened-heat treated components and includes a Delrin bearing type.
  • Designed for hard surfaces
  • Thermoplastic rubber wheel material
  • 6 in wheel diameter
  • 1-1/2 in wheel width
  • 280 lb load capacity
  • Hardened-heat treated construction
  • Delrin bearing type
  • Plate mount style
  • 7-3/8 in overall height
  • Nickel finish
Catalog No. 50-005-13261 Supplier MSC Supplier No. 01507623
This swivel top plate caster features a thermoplastic rubber wheel with a 6 in diameter and 1-1/2 in width. It offers a load capacity of 280 lb and an overall height of 7-3/8 in, making it suitable for various applications on hard surfaces. The caster is constructed with hardened-heat treated components and includes a Delrin bearing type.
  • Designed for hard surfaces
  • Thermoplastic rubber wheel material
  • 6 in wheel diameter
  • 1-1/2 in wheel width
  • 280 lb load capacity
  • Hardened-heat treated construction
  • Delrin bearing type
  • Plate mount style
  • 7-3/8 in overall height
  • Nickel finish