Learn More
Medchemexpress LLC E-Selectin/CD62E Mo 500ug
SDP
Supplier:  Medchemexpress LLC HYP73042500UG
Encompass_Preferred
E-Selectin/CD62E protein a cell-surface glycoprotein mediates the adhesion of blood neutrophils to cytokine-activated endothelium through interaction with SELPLG/PSGL1 It also potentially contributes to capillary morphogenesis and interacts with SELPLG/PSGL1 and PODXL2 These multifaceted interactions highlight E-Selectin s role in cell adhesion and its potential influence on vascular development E-Selectin/CD62E Protein Mouse (HEK293 His) is the recombinant mouse-derived E-Selectin/CD62E protein expressed by HEK293 with C-His C-10 His labeled tag
Catalog No. 50-001-81008