Learn More
Medchemexpress LLC E-Selectin/CD62E Ra 500ug
SDP
Catalog No. 5000181009
Encompass_Preferred
The E-selectin/CD62E protein is a cell surface glycoprotein that critically mediates immune adhesion by promoting neutrophil adhesion to cytokine-activated endothelial cells through the SELPLG/PSGL1 interaction It may also contribute to capillary morphogenesis and interact with SELPLG/PSGL1 and PODXL2 via the sialyl Lewis X epitope regardless of SELPLG sulfation E-Selectin/CD62E Protein Rat (HEK293 His) is the recombinant rat-derived E-Selectin/CD62E protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81009 Supplier Medchemexpress LLC Supplier No. HYP73044500UG
The E-selectin/CD62E protein is a cell surface glycoprotein that critically mediates immune adhesion by promoting neutrophil adhesion to cytokine-activated endothelial cells through the SELPLG/PSGL1 interaction It may also contribute to capillary morphogenesis and interact with SELPLG/PSGL1 and PODXL2 via the sialyl Lewis X epitope regardless of SELPLG sulfation E-Selectin/CD62E Protein Rat (HEK293 His) is the recombinant rat-derived E-Selectin/CD62E protein expressed by HEK293 with C-His labeled tag