Learn More
ACROBiosystems ACROBiosystems Human E-Selectin / CD62E Protein, His Tag
Catalog No. 501957463
Encompass_Preferred
Recombinant Protein;200UG;Human E-Selectin, His Tag (CDE-H5222) is expressed from human 293 cells (HEK293). It contains AA Trp 22 - Pro 556 (Accession # NP_000441.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.5 kDa. The protein migrates as 110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-7463 Supplier ACROBiosystems Supplier No. CDEH5222200UG
Recombinant Protein;200UG;Human E-Selectin, His Tag (CDE-H5222) is expressed from human 293 cells (HEK293). It contains AA Trp 22 - Pro 556 (Accession # NP_000441.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.5 kDa. The protein migrates as 110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.