Learn More
Sino Biological Human E-Selectin / SELE Protein (Fc Tag) 100ug
Supplier:  Sino Biological 10335H02H100
Encompass_Preferred
A DNA sequence encoding the human SELE (NP_000441.2) (Met1-Pro556) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-5060