Learn More
Invitrogen™ Human E-selectin (Soluble) ELISA Kit
Description
Specifications
Specifications
| Accession Number | P16581 |
| Assay Range | 1.6 to 50 ng/mL |
| Assay Sensitivity | 0.017 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 6401 |
| Gene Symbol | SELE |
| Interassay CV | 3.3% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.