Learn More
Cayman Chemical E-SelectInCD62Emousereco 100ug
SDP
Catalog No. 502910305
Encompass_Preferred
Source: Active recombinant C-terminal human IgG1 Fc-tagged mouse E-selectin expressed in HEK293 cells • Amino acids: 29-564 • MW: 85.5 kDa
Catalog No. 50-291-0305 Supplier Cayman Chemical Supplier No. 35726100 UG
Source: Active recombinant C-terminal human IgG1 Fc-tagged mouse E-selectin expressed in HEK293 cells • Amino acids: 29-564 • MW: 85.5 kDa