Description
- Secure seal between the oxygen e-tank and the yoke of the regulator
- Brass and Viton construction
- Robust, longer lasting seal
- Provided as a pack of 6
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Animal | All Animals |
| Type | Round Gasket |
| Disposable | Reusable |
| Size | Standard |
| Sterility | Non-sterile |