Learn More
Grainger E-Track 3 3/8 W 6 000 lb
Catalog No. 502766384
Encompass_Preferred
The Grainger website features a variety of truck trailer anchors and tie downs for securing cargo during transport. These products are designed to attach to truck trailers, flatbeds, and cargo containers to provide fixed anchor points. They are made from durable materials like galvanized steel and feature designs like D-rings, eye bolts, E-track fittings, and more. The anchors have weight ratings from 3,000 to 10,000 lbs and are available in different sizes and styles to accommodate light to heavy duty hauling needs. These cargo control products help prevent load shifting and damage during transport.
Catalog No. 50-276-6384 Supplier Grainger Supplier No. 16V903
The Grainger website features a variety of truck trailer anchors and tie downs for securing cargo during transport. These products are designed to attach to truck trailers, flatbeds, and cargo containers to provide fixed anchor points. They are made from durable materials like galvanized steel and feature designs like D-rings, eye bolts, E-track fittings, and more. The anchors have weight ratings from 3,000 to 10,000 lbs and are available in different sizes and styles to accommodate light to heavy duty hauling needs. These cargo control products help prevent load shifting and damage during transport.