Learn More
Grainger E-Track 4 7/8 W 6 000 lb
Catalog No. 502766362
Encompass_Preferred
The ANCRA Horizontal E-Track is a versatile metal slotted rail system that allows for adjustable mounting and positioning of hooks, hangers, clamps, stops and other accessories. It is made from extruded aluminum with a durable black anodized finish and features slots that run the length of the track to accept bolt on accessories. This horizontal E-Track measures 96" long x 1-5/8" wide x 1-3/8" high and can be mounted to walls, ceilings, floors, vehicles, machinery and more using the pre-drilled holes. It enables flexible organization, storage and material handling solutions.
Catalog No. 50-276-6362 Supplier Grainger Supplier No. 16V914
The ANCRA Horizontal E-Track is a versatile metal slotted rail system that allows for adjustable mounting and positioning of hooks, hangers, clamps, stops and other accessories. It is made from extruded aluminum with a durable black anodized finish and features slots that run the length of the track to accept bolt on accessories. This horizontal E-Track measures 96" long x 1-5/8" wide x 1-3/8" high and can be mounted to walls, ceilings, floors, vehicles, machinery and more using the pre-drilled holes. It enables flexible organization, storage and material handling solutions.