Learn More
Grainger E-Track 4 7/8 W 6 000 lb
Catalog No. 502766391
Encompass_Preferred
The Grainger 16V913 is a Dayton 1/3 HP Direct Drive Blower Motor that runs on 115V AC power at 1725 RPM. It has a 48 frame size, is single phase, and can provide airflow up to 850 CFM. The motor is thermally protected, has ball bearings, and is totally enclosed for protection against dust and moisture. It measures 9-1/2" long by 9-1/2" wide by 7-1/4" high. This direct drive motor is designed for air moving applications like ventilation systems, cooling towers, dust collection, and conveying systems.
Catalog No. 50-276-6391 Supplier Grainger Supplier No. 16V913
The Grainger 16V913 is a Dayton 1/3 HP Direct Drive Blower Motor that runs on 115V AC power at 1725 RPM. It has a 48 frame size, is single phase, and can provide airflow up to 850 CFM. The motor is thermally protected, has ball bearings, and is totally enclosed for protection against dust and moisture. It measures 9-1/2" long by 9-1/2" wide by 7-1/4" high. This direct drive motor is designed for air moving applications like ventilation systems, cooling towers, dust collection, and conveying systems.