Learn More
MSC Kinedyne Logistic Cam Strap, Polyester, 12 ft x 2 in., 835 Lb. Load Capacity, 1 ea
Catalog No. 5000547903
Encompass
This cargo control cam logistic strap is designed for securing items within transport vehicles using Series E or A track systems. The 12 ft. (3.66 m) strap is constructed from durable gold polyester webbing, featuring a spring-loaded end fitting for reliable attachment to vertical or horizontal tracks. It incorporates a cam buckle tensioning device for efficient load securement.
  • Working load limit of 835 lbs. (378.75 kg)
  • Ultimate load strength capacity of 2,505 lbs. (1,136.26 kg)
  • Equipped with FE8306-1 spring-loaded series E or A fitting
  • Features 910 cam buckle tensioning device
  • Webbing constructed from durable gold polyester
  • Suitable for use with logistic straps or beams
Catalog No. 50-005-47903 Supplier MSC Supplier No. 70904669
This cargo control cam logistic strap is designed for securing items within transport vehicles using Series E or A track systems. The 12 ft. (3.66 m) strap is constructed from durable gold polyester webbing, featuring a spring-loaded end fitting for reliable attachment to vertical or horizontal tracks. It incorporates a cam buckle tensioning device for efficient load securement.
  • Working load limit of 835 lbs. (378.75 kg)
  • Ultimate load strength capacity of 2,505 lbs. (1,136.26 kg)
  • Equipped with FE8306-1 spring-loaded series E or A fitting
  • Features 910 cam buckle tensioning device
  • Webbing constructed from durable gold polyester
  • Suitable for use with logistic straps or beams