Learn More
Medchemexpress LLC (E/Z)-4-Hydroxytamoxifen (Afimoxifene) | 68392-35-8 | 99.6% | 387.51 | 5 MG
SDP
Catalog No. 5000391306
Encompass_Preferred
(E/Z)-4-Hydroxytamoxifen (Afimoxifene) is a racemic compound consisting of (Z)-4-Hydroxytamoxifen and (E)-4-Hydroxytamoxifen isomers. It acts as a selective estrogen receptor modulator with both estrogenic and antiestrogenic activities and is an active metabolite of Tamoxifen.
  • Acts as a selective estrogen receptor modulator with both estrogenic and antiestrogenic activities.
  • Is an active metabolite of Tamoxifen.
  • Acts as an agonist of the G protein-coupled estrogen receptor (GPER) with relatively low affinity (100-1000 nM).
  • Shows promise for research into cyclical mastalgia, including breast pain, tenderness, and nodularity.
Catalog No. 50-003-91306 Supplier Medchemexpress LLC Supplier No. HY16950A5MG
(E/Z)-4-Hydroxytamoxifen (Afimoxifene) is a racemic compound consisting of (Z)-4-Hydroxytamoxifen and (E)-4-Hydroxytamoxifen isomers. It acts as a selective estrogen receptor modulator with both estrogenic and antiestrogenic activities and is an active metabolite of Tamoxifen.
  • Acts as a selective estrogen receptor modulator with both estrogenic and antiestrogenic activities.
  • Is an active metabolite of Tamoxifen.
  • Acts as an agonist of the G protein-coupled estrogen receptor (GPER) with relatively low affinity (100-1000 nM).
  • Shows promise for research into cyclical mastalgia, including breast pain, tenderness, and nodularity.