Learn More
Omega Biotek Inc E-Z 96 DNA PLATES,10 PREPS
Catalog No. NC1467247
Encompass
E-Z 96® DNA Plates, 10 prepsE-Z 96® plates are silica glass fiber plates that can bind up to 50 µg of genomic DNA or 20 µg of plasmid DNA per prep. Each well has a capacity of 1 mL and the plates are semi-skirted. E-Z 96® DNA plates can be processed in vacuum manifolds and/or by centrifugation. Omega Bio-tek columns are compatible with following Qiagen Kits: Qiaprep Spin Mini, QiaQuick Gel Extraction, QiaQuick PCR Clean-up, Dneasy Blood & Tissue, DNeasy Plant DNA.
Catalog No. NC1467247 Supplier Omega Biotek Inc Supplier No. BD9601
Add to Cart
Add to Cart
E-Z 96® DNA Plates, 10 prepsE-Z 96® plates are silica glass fiber plates that can bind up to 50 µg of genomic DNA or 20 µg of plasmid DNA per prep. Each well has a capacity of 1 mL and the plates are semi-skirted. E-Z 96® DNA plates can be processed in vacuum manifolds and/or by centrifugation. Omega Bio-tek columns are compatible with following Qiagen Kits: Qiaprep Spin Mini, QiaQuick Gel Extraction, QiaQuick PCR Clean-up, Dneasy Blood & Tissue, DNeasy Plant DNA.