Learn More
Omega Biotek Inc E-Z 96 Total RNA Kit, 12 x 96 Preps, 96-well plate format, HiBind silica technology, RNA purification, processing time < 60 min, RNase-free water elution
Catalog No. NC1837579
Encompass
E-Z 96 Total RNA Kit, 12 x 96 Preps, 96-well plate format, HiBind silica technology, RNA purification, processing time < 60 min, RNase-free water elution
Catalog No. NC1837579 Supplier Omega Biotek Inc Supplier No. R103402
E-Z 96 Total RNA Kit, 12 x 96 Preps, 96-well plate format, HiBind silica technology, RNA purification, processing time < 60 min, RNase-free water elution