Learn More
Medchemexpress LLC (E/Z)-Afatinib | 439081-18-2 | MFCD12407405 | 100.0% | 485.94 | C24H25ClFN5O3 | 1 ML
SDP
Catalog No. 5000245530
Encompass_Preferred
(E/Z)-Afatinib is a research-grade mixture of the E and Z isomers of Afatinib, an irreversible inhibitor of epidermal growth factor receptor (EGFR) family kinases. It is intended for laboratory research use and is supplied as a concentrated solution for biochemical and cell-based assays.
  • Irreversibly inhibits EGFR, HER2, and HER4.
  • Mixture of E and Z isomers for comprehensive activity profiling.
  • Provided as a 10 mM solution in DMSO, 1 mL, suitable for stock preparation.
  • High purity (99.98% by HPLC) suitable for biochemical assays.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-45530 Supplier Medchemexpress LLC Supplier No. HY10261B10MM 1ML
(E/Z)-Afatinib is a research-grade mixture of the E and Z isomers of Afatinib, an irreversible inhibitor of epidermal growth factor receptor (EGFR) family kinases. It is intended for laboratory research use and is supplied as a concentrated solution for biochemical and cell-based assays.
  • Irreversibly inhibits EGFR, HER2, and HER4.
  • Mixture of E and Z isomers for comprehensive activity profiling.
  • Provided as a 10 mM solution in DMSO, 1 mL, suitable for stock preparation.
  • High purity (99.98% by HPLC) suitable for biochemical assays.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet.