Learn More
Medchemexpress LLC (E/Z)-AG490 | 134036-52-5 | MFCD00236452 | 99.7% | 294.30 g/mol | C17H14N2O3 | 5 MG
SDP
Catalog No. 5000239887
Encompass_Preferred
(E/Z)-AG490 is a racemic mixture of the (E)- and (Z)-isomers of AG490 supplied as a solid research reagent. The (E)-isomer is a tyrosine kinase inhibitor reported to inhibit EGFR, STAT3, and JAK2/3; the compound is used in biochemical and cell-based studies to probe kinase-dependent signaling pathways. High-purity material is provided for laboratory research.
Catalog No. 50-002-39887 Supplier Medchemexpress LLC Supplier No. HY1074595MG
(E/Z)-AG490 is a racemic mixture of the (E)- and (Z)-isomers of AG490 supplied as a solid research reagent. The (E)-isomer is a tyrosine kinase inhibitor reported to inhibit EGFR, STAT3, and JAK2/3; the compound is used in biochemical and cell-based studies to probe kinase-dependent signaling pathways. High-purity material is provided for laboratory research.