Learn More
Medchemexpress LLC E/Z-C20 ceramide (N-arachidoyl-D-sphingosine) | 123482-93-9 | MFCD02181210 | 99.9% | 594.01 | C38H75NO3 | 5 MG
SDP
Catalog No. 5000264732
Encompass_Preferred
(E/Z)-C20 Ceramide is a research-grade mixture of the E- and Z-isomers of N-arachidoyl-D-sphingosine (a 20:0 ceramide). CAS 123482-93-9; molecular formula C38H75NO3; molecular weight 594.01. Supplied as a solid for biochemical and cell biology research involving sphingolipid metabolism and signaling.
  • High purity (~99.9%) suitable for analytical and biological studies.
  • White to off-white solid appearance.
  • Stable as powder at -20°C for up to 3 years; in solvent store at -80°C for up to 6 months or -20°C for 1 month.
  • Mixture of E and Z isomers relevant to sphingolipid research.
  • For research use only; not for human or therapeutic use.
  • Datasheet and certificate of analysis available for quality verification.
Catalog No. 50-002-64732 Supplier Medchemexpress LLC Supplier No. HY112016A5MG
(E/Z)-C20 Ceramide is a research-grade mixture of the E- and Z-isomers of N-arachidoyl-D-sphingosine (a 20:0 ceramide). CAS 123482-93-9; molecular formula C38H75NO3; molecular weight 594.01. Supplied as a solid for biochemical and cell biology research involving sphingolipid metabolism and signaling.
  • High purity (~99.9%) suitable for analytical and biological studies.
  • White to off-white solid appearance.
  • Stable as powder at -20°C for up to 3 years; in solvent store at -80°C for up to 6 months or -20°C for 1 month.
  • Mixture of E and Z isomers relevant to sphingolipid research.
  • For research use only; not for human or therapeutic use.
  • Datasheet and certificate of analysis available for quality verification.