Learn More
Medchemexpress LLC (E/Z)-FK-506 26,28-allylic ester rearrangement impurity | 131944-48-4 | 804.02 | C44H69NO12 | 5 MG
SDP
Catalog No. 5000284913
Encompass_Preferred
(E/Z)-FK-506 26,28-allylic ester rearrangement impurity (FK506-A) is an impurity of tacrolimus supplied as a reference substance for analytical and research applications. It is intended for use in impurity identification, method development, and stability studies where characterization of tacrolimus-related degradation or synthesis by-products is required.
  • Provides a characterized impurity reference for analytical validation.
  • Enables impurity profiling and stability testing of tacrolimus samples.
  • Compatible with HPLC and LC-MS analytical workflows.
  • Offered in small milligram quantities suitable for method development.
  • Includes CAS and molecular data to support documentation.
Catalog No. 50-002-84913 Supplier Medchemexpress LLC Supplier No. HYW5858865MG
(E/Z)-FK-506 26,28-allylic ester rearrangement impurity (FK506-A) is an impurity of tacrolimus supplied as a reference substance for analytical and research applications. It is intended for use in impurity identification, method development, and stability studies where characterization of tacrolimus-related degradation or synthesis by-products is required.
  • Provides a characterized impurity reference for analytical validation.
  • Enables impurity profiling and stability testing of tacrolimus samples.
  • Compatible with HPLC and LC-MS analytical workflows.
  • Offered in small milligram quantities suitable for method development.
  • Includes CAS and molecular data to support documentation.