Learn More
Medchemexpress LLC E/Z-flupentixol dihydrochloride-d4 | 1246833-30-6 | >95.0% | 438.54 g/mol | C23H21D4F3N2OS | 1 MG
SDP
Catalog No. 5000283922
Encompass_Preferred
(E/Z)-Flupentixol dihydrochloride-d4 is a deuterium-labeled isotopic analogue of (E/Z)-flupentixol dihydrochloride supplied as a 1 mg research-grade standard. It is intended for use as an internal standard or tracer in quantitative analytical methods, including NMR, LC-MS, and GC-MS, and for studies of drug metabolism and pharmacokinetics. The product is for research use only.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, LC-MS, and GC-MS applications.
  • Salt form: dihydrochloride; labeled as d4.
  • Supplied as a 1 mg research sample.
  • Useful for pharmacokinetic and drug metabolism studies.
  • Store under conditions specified in the certificate of analysis; purity typically >95% by HPLC.
Catalog No. 50-002-83922 Supplier Medchemexpress LLC Supplier No. HYW6541971MG
(E/Z)-Flupentixol dihydrochloride-d4 is a deuterium-labeled isotopic analogue of (E/Z)-flupentixol dihydrochloride supplied as a 1 mg research-grade standard. It is intended for use as an internal standard or tracer in quantitative analytical methods, including NMR, LC-MS, and GC-MS, and for studies of drug metabolism and pharmacokinetics. The product is for research use only.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, LC-MS, and GC-MS applications.
  • Salt form: dihydrochloride; labeled as d4.
  • Supplied as a 1 mg research sample.
  • Useful for pharmacokinetic and drug metabolism studies.
  • Store under conditions specified in the certificate of analysis; purity typically >95% by HPLC.