Learn More
Medchemexpress LLC (E/Z)-Ginkgolic acid C17:2 | 102811-39-2 | 1 MG

Supplier: Medchemexpress LLC HYN79611MG
(E/Z)-Ginkgolic acid C17:2, isolated from Ginkgo biloba, can bind tightly with human dihydroorotate dehydrogenase (DHODH). It is for research use only and not sold to patients.
- Purity: 99.37%
- Appearance: Oil
- Color: Off-white to light yellow
- Initial source: Plants, Ginkgoaceae, Ginkgo biloba
- Molecular weight: 372.54
- Formula: C24H36O3
- SMILES: O=C(O)C1=C(O)C=CC=C1CCCCCCC/C=C/C/C=C/CCCCC
- Structure classification: Phenols, Monophenols, Ketones, Aldehydes, Acids
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.