Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E/Z)-Ginkgolic acid C17:2 | 102811-39-2 | MFCD30609542 | 5 MG
SDP

Catalog No. 5000417577
Encompass_Preferred

(E/Z)-Ginkgolic acid C17:2 is a compound isolated from *Ginkgo biloba*. It has demonstrated the ability to bind tightly with human dihydroorotate dehydrogenase (DHODH).

  • Isolated from *Ginkgo biloba*.
  • Binds with human dihydroorotate dehydrogenase (DHODH).
  • Purity: 99.37%.
  • Appearance: off-white to light yellow oil.
  • Molecular weight: 372.54.
  • Formula: C24H36O3.
  • CAS no.: 102811-39-2.

Catalog No. 50-004-17577 Supplier Medchemexpress LLC Supplier No. HYN79615MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

(E/Z)-Ginkgolic acid C17:2 is a compound isolated from *Ginkgo biloba*. It has demonstrated the ability to bind tightly with human dihydroorotate dehydrogenase (DHODH).

  • Isolated from *Ginkgo biloba*.
  • Binds with human dihydroorotate dehydrogenase (DHODH).
  • Purity: 99.37%.
  • Appearance: off-white to light yellow oil.
  • Molecular weight: 372.54.
  • Formula: C24H36O3.
  • CAS no.: 102811-39-2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.