Learn More
Medchemexpress LLC (E/Z)-GSK5182 | 2699724-40-6 | 98.9% | 417.54 | 50 MG
SDP
Catalog No. 5000343208
Encompass_Preferred
(E/Z)-GSK5182 is a racemic compound of (E)-GSK5182 and (Z)-GSK5182 isomers. It is a highly selective and orally active inverse agonist of estrogen-related receptor γ (ERRγ) with an IC50 of 79 nM. It also induces reactive oxygen species (ROS) generation in hepatocellular carcinoma.
  • Racemic compound.
  • Highly selective and orally active.
  • Inverse agonist of estrogen-related receptor γ (ERRγ) with an IC50 of 79 nM.
  • Induces reactive oxygen species (ROS) generation in hepatocellular carcinoma.
Catalog No. 50-003-43208 Supplier Medchemexpress LLC Supplier No. HY111226A10MM/1ML
(E/Z)-GSK5182 is a racemic compound of (E)-GSK5182 and (Z)-GSK5182 isomers. It is a highly selective and orally active inverse agonist of estrogen-related receptor γ (ERRγ) with an IC50 of 79 nM. It also induces reactive oxygen species (ROS) generation in hepatocellular carcinoma.
  • Racemic compound.
  • Highly selective and orally active.
  • Inverse agonist of estrogen-related receptor γ (ERRγ) with an IC50 of 79 nM.
  • Induces reactive oxygen species (ROS) generation in hepatocellular carcinoma.