Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC E/Z-HA155 100mg | 1229652-22-5 | 463.29 g/mol | C24H19BFNO5S | 100 MG
SDP

Catalog No. 5000229780
Encompass_Preferred

(E/Z)-HA155 is a potent autotaxin (ATX) type I inhibitor used as a research reagent in studies of cancer, fibrotic diseases, inflammation, pain, and angiogenesis. The compound is provided as solid and as DMSO solutions, has high reported purity, and specific storage conditions are supplied for long-term stability.

  • Potent autotaxin (ATX) type I inhibitor for biochemical and cellular assays.
  • High reported purity: 99.26%.
  • Available in multiple quantities including 100 MG solid and 10 mM solution in DMSO.
  • Chemical formula C24H19BFNO5S and molecular weight 463.29 g/mol for formulation reference.
  • Storage recommendations: powder at -20°C for up to 3 years; in solvent at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-29780 Supplier Medchemexpress LLC Supplier No. HY116100100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E/Z)-HA155 is a potent autotaxin (ATX) type I inhibitor used as a research reagent in studies of cancer, fibrotic diseases, inflammation, pain, and angiogenesis. The compound is provided as solid and as DMSO solutions, has high reported purity, and specific storage conditions are supplied for long-term stability.

  • Potent autotaxin (ATX) type I inhibitor for biochemical and cellular assays.
  • High reported purity: 99.26%.
  • Available in multiple quantities including 100 MG solid and 10 mM solution in DMSO.
  • Chemical formula C24H19BFNO5S and molecular weight 463.29 g/mol for formulation reference.
  • Storage recommendations: powder at -20°C for up to 3 years; in solvent at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.