Learn More
Medchemexpress LLC E/Z-HA155 1mg | 1229652-22-5 | 463.29 g·mol-1 | C24H19BFNO5S | 1 MG
SDP
Catalog No. 5000227931
Encompass_Preferred
(E/Z)-HA155 is a boronic acid-based autotaxin (ATX) type I inhibitor used in preclinical research. It is reported to inhibit ATX with high potency (IC50 ≈ 5.7 nM) and is used to study cancer, fibrotic diseases, inflammation, pain, and angiogenesis.
  • Potent autotaxin (ATX) type I inhibitor (IC50 ≈ 5.7 nM).
  • High purity: 99.26%.
  • Available in multiple lab-scale pack sizes, including 1 mg vials and DMSO solution.
  • Molecular formula C24H19BFNO5S; molecular weight 463.29 g·mol-1.
  • Supplied as an E/Z isomer mixture suitable for biochemical and cell-based assays.
Catalog No. 50-002-27931 Supplier Medchemexpress LLC Supplier No. HY1161001MG
(E/Z)-HA155 is a boronic acid-based autotaxin (ATX) type I inhibitor used in preclinical research. It is reported to inhibit ATX with high potency (IC50 ≈ 5.7 nM) and is used to study cancer, fibrotic diseases, inflammation, pain, and angiogenesis.
  • Potent autotaxin (ATX) type I inhibitor (IC50 ≈ 5.7 nM).
  • High purity: 99.26%.
  • Available in multiple lab-scale pack sizes, including 1 mg vials and DMSO solution.
  • Molecular formula C24H19BFNO5S; molecular weight 463.29 g·mol-1.
  • Supplied as an E/Z isomer mixture suitable for biochemical and cell-based assays.