Learn More
Medchemexpress LLC E/Z-HA155 25mg | 1229652-22-5 | 463.29 | C24H19BFNO5S | 25 MG
SDP
Catalog No. 5000228949
Encompass_Preferred
A potent autotaxin (ATX) type I inhibitor for laboratory research, (E/Z)-HA155 is used to study cancer, fibrotic disease, inflammation, pain, and angiogenesis. The product is supplied as a high-purity solid and includes supporting structural and stability data for experimental use.
  • Potent autotaxin (ATX) type I inhibitor for mechanistic and pharmacology studies.
  • High purity suitable for analytical and biological assays.
  • Supplied as a solid 25 mg quantity for small-scale experiments.
  • Stable when stored as a powder at -20°C for extended periods.
  • Structural identifiers and formula provided for computational and analytical workflows.
Catalog No. 50-002-28949 Supplier Medchemexpress LLC Supplier No. HY11610025MG
A potent autotaxin (ATX) type I inhibitor for laboratory research, (E/Z)-HA155 is used to study cancer, fibrotic disease, inflammation, pain, and angiogenesis. The product is supplied as a high-purity solid and includes supporting structural and stability data for experimental use.
  • Potent autotaxin (ATX) type I inhibitor for mechanistic and pharmacology studies.
  • High purity suitable for analytical and biological assays.
  • Supplied as a solid 25 mg quantity for small-scale experiments.
  • Stable when stored as a powder at -20°C for extended periods.
  • Structural identifiers and formula provided for computational and analytical workflows.