Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC E/Z-HA155 50mg | 1229652-22-5 | 50 MG
SDP

Catalog No. 5000227933
Encompass_Preferred

(E/Z)-HA155 is a boronic acid small-molecule autotaxin (ATX) type I inhibitor supplied as an E/Z isomer mixture for preclinical research. It is used to study cancer, fibrotic diseases, inflammation, pain, and angiogenesis. Supplied as a high-purity solid suitable for biochemical and cell-based assays; recommended storage is powder at -20°C.

  • Boronic acid small molecule inhibitor of autotaxin.
  • Used in research on cancer, fibrosis, inflammation, pain, and angiogenesis.
  • Molecular formula C24H19BFNO5S.
  • Molecular weight 463.29 g/mol.
  • Purity approximately 99.3%.
  • Appearance: solid, off-white to light yellow.
  • Recommended storage: powder at -20°C; in solvent keep at -80°C for longer-term, -20°C short-term.

Catalog No. 50-002-27933 Supplier Medchemexpress LLC Supplier No. HY11610050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E/Z)-HA155 is a boronic acid small-molecule autotaxin (ATX) type I inhibitor supplied as an E/Z isomer mixture for preclinical research. It is used to study cancer, fibrotic diseases, inflammation, pain, and angiogenesis. Supplied as a high-purity solid suitable for biochemical and cell-based assays; recommended storage is powder at -20°C.

  • Boronic acid small molecule inhibitor of autotaxin.
  • Used in research on cancer, fibrosis, inflammation, pain, and angiogenesis.
  • Molecular formula C24H19BFNO5S.
  • Molecular weight 463.29 g/mol.
  • Purity approximately 99.3%.
  • Appearance: solid, off-white to light yellow.
  • Recommended storage: powder at -20°C; in solvent keep at -80°C for longer-term, -20°C short-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.