Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC E/Z-HA155 5mg | 1229652-22-5 | 5 MG
SDP

Catalog No. 5000227275
Encompass_Preferred

(E/Z)-HA155 is a small-molecule autotaxin (ATX) type I inhibitor used in preclinical research to study cancer, fibrotic diseases, inflammation, pain, and angiogenesis. Supplied as a high-purity powder with recommended cold storage, it is available in multiple pack sizes and as a DMSO solution for biological assays.

  • High purity: 99.3% (rounded from manufacturer specification).
  • Molecular weight approximately 463.29 g·mol⁻¹.
  • Chemical formula C24H19BFNO5S.
  • Recommended storage: powder at -20°C; solutions at -80°C for long-term storage.
  • Available in small research quantities and as a 10 mM solution in DMSO for assay use.
  • Supplied with a certificate of analysis and safety data sheet for quality and handling information.

Catalog No. 50-002-27275 Supplier Medchemexpress LLC Supplier No. HY1161005MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E/Z)-HA155 is a small-molecule autotaxin (ATX) type I inhibitor used in preclinical research to study cancer, fibrotic diseases, inflammation, pain, and angiogenesis. Supplied as a high-purity powder with recommended cold storage, it is available in multiple pack sizes and as a DMSO solution for biological assays.

  • High purity: 99.3% (rounded from manufacturer specification).
  • Molecular weight approximately 463.29 g·mol⁻¹.
  • Chemical formula C24H19BFNO5S.
  • Recommended storage: powder at -20°C; solutions at -80°C for long-term storage.
  • Available in small research quantities and as a 10 mM solution in DMSO for assay use.
  • Supplied with a certificate of analysis and safety data sheet for quality and handling information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.