Learn More
Euthanex EZ-1000 E-Z HEAT Controller with temperature range of 95-104 degrees Fahrenheit (Heated Beds sold separately).
Catalog No. NC1880358
Encompass_Preferred
EZ-1000 E-Z Heat is an extremely user-friendly system that allows up to four connected beds to help maintain the patients" body temperatures during surgical procedures. This patented warming system allows the user to heat the induction chamber and warming beds simultaneously. The warming beds ranges from 95 to 104 degrees Fahrenheit. Bed temperatures can be adjusted in the administrator options. Each of the warming circuits displays the elapsed time and the set temperature, which can be set individually. All settings made in the administrator options are password-protected to ensure total security from tampering individuals. The heating bed also allows the option to mount our Microflex Breather and works in perfect harmony with all E-Z Anesthesia systems."
Catalog No. NC1880358 Supplier Euthanex Supplier No. EZ1000
EZ-1000 E-Z Heat is an extremely user-friendly system that allows up to four connected beds to help maintain the patients" body temperatures during surgical procedures. This patented warming system allows the user to heat the induction chamber and warming beds simultaneously. The warming beds ranges from 95 to 104 degrees Fahrenheit. Bed temperatures can be adjusted in the administrator options. Each of the warming circuits displays the elapsed time and the set temperature, which can be set individually. All settings made in the administrator options are password-protected to ensure total security from tampering individuals. The heating bed also allows the option to mount our Microflex Breather and works in perfect harmony with all E-Z Anesthesia systems."