Learn More
Medchemexpress LLC (E/Z)-Icerguastat | 13098-73-2 | 99.4% | 196.64 | 5 MG
SDP
Catalog No. 5000385760
Encompass_Preferred
(E/Z)-Icerguastat is a selective inhibitor of the phosphatase regulatory subunit PPP1R15A (R15A). It is designed for protein misfolding diseases research.
  • Selective inhibitor of the phosphatase regulatory subunit PPP1R15A (R15A)
  • Can be used for protein misfolding diseases research
  • Target is Phosphatase
  • Pathway is Metabolic Enzyme/Protease
Catalog No. 50-003-85760 Supplier Medchemexpress LLC Supplier No. HY189565MG
(E/Z)-Icerguastat is a selective inhibitor of the phosphatase regulatory subunit PPP1R15A (R15A). It is designed for protein misfolding diseases research.
  • Selective inhibitor of the phosphatase regulatory subunit PPP1R15A (R15A)
  • Can be used for protein misfolding diseases research
  • Target is Phosphatase
  • Pathway is Metabolic Enzyme/Protease