Learn More
Selleck Chemical LLC (E/Z)-IT-603 S3384-5mg
SDP
Catalog No. 503108329
Encompass_Preferred
(E/Z)-IT-603 is a mixture of E-IT-603 and Z-IT-603 IT-603 is a NF- B family member c-Rel inhibitor with an IC50 of 3 M in electrophoretic mobility shift assay (EMSA)
Catalog No. 50-310-8329 Supplier Selleck Chemical LLC Supplier No. S33845MG
(E/Z)-IT-603 is a mixture of E-IT-603 and Z-IT-603 IT-603 is a NF- B family member c-Rel inhibitor with an IC50 of 3 M in electrophoretic mobility shift assay (EMSA)