Learn More
Medchemexpress LLC (E/Z)-J147 | 1146963-51-0 | 99.5% | 350.34 | 1 ML
SDP
Catalog No. 5000350960
Encompass_Preferred
(E/Z)-J147 is a potent, orally active, neuroprotective agent designed for cognitive enhancement. It readily crosses the blood-brain barrier and is being studied for its potential in treating Alzheimer's disease. The compound can inhibit monoamine oxidase B and the dopamine transporter, promoting cell survival.
  • Exceptionally potent and orally active
  • Neuroprotective agent for cognitive enhancement
  • Readily crosses the blood-brain barrier
  • Inhibits monoamine oxidase B (MAO B) and the dopamine transporter
  • Promotes HT22 and primary cell survival
  • Enhances memory and dendritic spine number in old mice
  • Potential for Alzheimer's disease treatment
Catalog No. 50-003-50960 Supplier Medchemexpress LLC Supplier No. HY1377910MM/1ML
(E/Z)-J147 is a potent, orally active, neuroprotective agent designed for cognitive enhancement. It readily crosses the blood-brain barrier and is being studied for its potential in treating Alzheimer's disease. The compound can inhibit monoamine oxidase B and the dopamine transporter, promoting cell survival.
  • Exceptionally potent and orally active
  • Neuroprotective agent for cognitive enhancement
  • Readily crosses the blood-brain barrier
  • Inhibits monoamine oxidase B (MAO B) and the dopamine transporter
  • Promotes HT22 and primary cell survival
  • Enhances memory and dendritic spine number in old mice
  • Potential for Alzheimer's disease treatment