Learn More
Medchemexpress LLC (E/Z)-Locostatin | 133812-16-5 | 99.5% | 245.27 | 10 MG
SDP
Catalog No. 5000454681
Encompass_Preferred
(E/Z)-Locostatin, also known as (E/Z)-UIC-1005, is a racemic form of Locostatin. This potent RKIP inhibitor works by binding to the Raf kinase inhibitor RKIP protein, thereby disrupting its interaction with Raf-1 kinase and G protein-coupled receptor kinase 2. It has been shown to inhibit cell proliferation and migration. It can also aggravate thioacetamide-induced acute liver failure in mice.
  • Potent RKIP inhibitor
  • Inhibits cell proliferation and migration
  • Disrupts interaction of RKIP with Raf-1 kinase
  • Disrupts interaction of RKIP with G protein-coupled receptor kinase 2
Catalog No. 50-004-54681 Supplier Medchemexpress LLC Supplier No. HYW01341110MG
(E/Z)-Locostatin, also known as (E/Z)-UIC-1005, is a racemic form of Locostatin. This potent RKIP inhibitor works by binding to the Raf kinase inhibitor RKIP protein, thereby disrupting its interaction with Raf-1 kinase and G protein-coupled receptor kinase 2. It has been shown to inhibit cell proliferation and migration. It can also aggravate thioacetamide-induced acute liver failure in mice.
  • Potent RKIP inhibitor
  • Inhibits cell proliferation and migration
  • Disrupts interaction of RKIP with Raf-1 kinase
  • Disrupts interaction of RKIP with G protein-coupled receptor kinase 2