Learn More
Medchemexpress LLC (E/Z)-mirin | 299953-00-7 | 99.2% | 220.25 | C10H8N2O2S | 10 MG
SDP
Catalog No. 5000227076
Encompass_Preferred
(E/Z)-Mirin is a research-grade small molecule mixture of E and Z isomers that inhibits the MRN (Mre11-Rad50-Nbs1) complex and prevents MRN-dependent activation of ATM. It is intended for laboratory research use, and is provided with standard analytical documentation and storage recommendations.
  • Inhibits the MRN (Mre11-Rad50-Nbs1) complex and prevents MRN-dependent activation of ATM.
  • Research-grade mixture of E and Z isomers supplied for laboratory use only.
  • High purity, 99.22% (as provided by manufacturer).
  • Available as 10 mg solid and 10 mM solutions in DMSO for flexible dosing.
  • Characterized by COA, HNMR, RP-HPLC, and LCMS documentation.
  • Store sealed, away from moisture and light; in solvent store at -80°C for long-term storage.
Catalog No. 50-002-27076 Supplier Medchemexpress LLC Supplier No. HY11769310MG
(E/Z)-Mirin is a research-grade small molecule mixture of E and Z isomers that inhibits the MRN (Mre11-Rad50-Nbs1) complex and prevents MRN-dependent activation of ATM. It is intended for laboratory research use, and is provided with standard analytical documentation and storage recommendations.
  • Inhibits the MRN (Mre11-Rad50-Nbs1) complex and prevents MRN-dependent activation of ATM.
  • Research-grade mixture of E and Z isomers supplied for laboratory use only.
  • High purity, 99.22% (as provided by manufacturer).
  • Available as 10 mg solid and 10 mM solutions in DMSO for flexible dosing.
  • Characterized by COA, HNMR, RP-HPLC, and LCMS documentation.
  • Store sealed, away from moisture and light; in solvent store at -80°C for long-term storage.