Learn More
Omega Biotek Inc E.Z.N.A Plasmid Mini Kit I, 200 preps, 1-5 mL bacterial culture, yield up to 30 μg DNA, elution volume 50 μL, HiBind Mini Column, V-spin, manual processing, safety feature: no phenol/chloroform
Catalog No. NC0919496
Encompass
E.Z.N.A Plasmid Mini Kit I, 200 preps, 1-5 mL bacterial culture, yield up to 30 μg DNA, elution volume 50 μL, HiBind Mini Column, V-spin, manual processing, safety feature: no phenol/chloroform
Catalog No. NC0919496 Supplier Omega Biotek Inc Supplier No. D694302
E.Z.N.A Plasmid Mini Kit I, 200 preps, 1-5 mL bacterial culture, yield up to 30 μg DNA, elution volume 50 μL, HiBind Mini Column, V-spin, manual processing, safety feature: no phenol/chloroform