Learn More
Medchemexpress LLC (E/Z)-N-benzyl-2-cyano-3-(3,4-dihydroxyphenyl)prop-2-enamide | 134036-52-5 | MFCD00236452 | 99.7% | 294.30 g/mol | C17H14N2O3 | 10 MG
SDP
Catalog No. 5000241040
Encompass_Preferred
(E/Z)-AG490 is the racemic mixture of the (E) and (Z) isomers of AG490, a small-molecule tyrosine kinase inhibitor used in research to probe EGFR and JAK/STAT signaling. It is supplied as a high-purity solid and is commonly prepared as a DMSO solution for cellular and biochemical assays.
  • Racemic mixture of (E) and (Z) isomers for comparative signaling studies.
  • Inhibits EGFR, JAK2/3, and STAT3 pathways in biochemical and cell-based assays.
  • High purity (≈99.7%), suitable for research applications.
  • Solid powder form with recommended storage at -20°C; solutions stable at -80°C for long term.
  • Soluble in DMSO and commonly prepared as 10 mM solutions for in vitro testing.
Catalog No. 50-002-41040 Supplier Medchemexpress LLC Supplier No. HY10745910MG
(E/Z)-AG490 is the racemic mixture of the (E) and (Z) isomers of AG490, a small-molecule tyrosine kinase inhibitor used in research to probe EGFR and JAK/STAT signaling. It is supplied as a high-purity solid and is commonly prepared as a DMSO solution for cellular and biochemical assays.
  • Racemic mixture of (E) and (Z) isomers for comparative signaling studies.
  • Inhibits EGFR, JAK2/3, and STAT3 pathways in biochemical and cell-based assays.
  • High purity (≈99.7%), suitable for research applications.
  • Solid powder form with recommended storage at -20°C; solutions stable at -80°C for long term.
  • Soluble in DMSO and commonly prepared as 10 mM solutions for in vitro testing.