Learn More
Medchemexpress LLC E/Z-necrosulfonamide | 432531-71-0 | 98.8% | 461.47 g·mol⁻¹ | C18H15N5O6S2 | 100 MG
SDP
Catalog No. 5000211581
Encompass_Preferred
(E/Z)-Necrosulfonamide is a racemic small-molecule inhibitor of mixed lineage kinase domain-like protein (MLKL) and gasdermin D (GSDMD), used in cellular research to probe necroptosis and pyroptosis pathways. Supplied as a solid powder with high reported purity, the compound is provided in multiple package sizes and requires cold storage for long-term stability.
  • Inhibits MLKL- and GSDMD-mediated cell death.
  • High reported purity suitable for research assays.
  • Available in multiple package sizes for experimental flexibility.
  • Solid powder form simplifies handling and storage.
  • Stable under recommended refrigerated or frozen storage.
Catalog No. 50-002-11581 Supplier Medchemexpress LLC Supplier No. HY100573A100MG
(E/Z)-Necrosulfonamide is a racemic small-molecule inhibitor of mixed lineage kinase domain-like protein (MLKL) and gasdermin D (GSDMD), used in cellular research to probe necroptosis and pyroptosis pathways. Supplied as a solid powder with high reported purity, the compound is provided in multiple package sizes and requires cold storage for long-term stability.
  • Inhibits MLKL- and GSDMD-mediated cell death.
  • High reported purity suitable for research assays.
  • Available in multiple package sizes for experimental flexibility.
  • Solid powder form simplifies handling and storage.
  • Stable under recommended refrigerated or frozen storage.