Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E/Z)-necrosulfonamide | 432531-71-0 | MFCD02370191 | 98.8% | 461.47 | C18H15N5O6S2 | 10 MG
SDP

Catalog No. 5000212349
Encompass_Preferred

(E/Z)-Necrosulfonamide is a racemic small-molecule inhibitor used to study programmed cell death pathways, including necroptosis and pyroptosis. The compound has formula C18H15N5O6S2, molecular weight 461.47, and CAS 432531-71-0.

  • Racemic small-molecule inhibitor for cell death pathway studies.
  • Active against MLKL and gasdermin D.
  • Provided in a 10 mg research quantity.
  • High purity (~98.8% by HPLC), suitable for biochemical assays.
  • Molecular formula C18H15N5O6S2 and molecular weight 461.47.

Catalog No. 50-002-12349 Supplier Medchemexpress LLC Supplier No. HY100573A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E/Z)-Necrosulfonamide is a racemic small-molecule inhibitor used to study programmed cell death pathways, including necroptosis and pyroptosis. The compound has formula C18H15N5O6S2, molecular weight 461.47, and CAS 432531-71-0.

  • Racemic small-molecule inhibitor for cell death pathway studies.
  • Active against MLKL and gasdermin D.
  • Provided in a 10 mg research quantity.
  • High purity (~98.8% by HPLC), suitable for biochemical assays.
  • Molecular formula C18H15N5O6S2 and molecular weight 461.47.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.