Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC E/Z-necrosulfonamide | 432531-71-0 | MFCD02370191 | 98.8% | 461.47 | C18H15N5O6S2 | 50 MG
SDP

Catalog No. 5000211582
Encompass_Preferred

E/Z-necrosulfonamide is the racemic form of necrosulfonamide, a small-molecule inhibitor that targets the execution steps of necroptosis and pyroptosis. It is used in cellular and biochemical research to study MLKL- and gasdermin D-mediated necrotic pathways.

  • Racemic small-molecule inhibitor of mixed lineage kinase domain-like protein (MLKL) and gasdermin D (GSDMD).
  • Interferes with executor oligomerization to inhibit necroptosis and pyroptosis.
  • Suitable for research on neurodegeneration, ischemia-reperfusion injury, and inflammatory disorders.
  • Reported molecular formula C18H15N5O6S2 and molecular weight 461.47.
  • High reported purity (98.8%) and multiple package sizes for experimental flexibility.

Catalog No. 50-002-11582 Supplier Medchemexpress LLC Supplier No. HY100573A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

E/Z-necrosulfonamide is the racemic form of necrosulfonamide, a small-molecule inhibitor that targets the execution steps of necroptosis and pyroptosis. It is used in cellular and biochemical research to study MLKL- and gasdermin D-mediated necrotic pathways.

  • Racemic small-molecule inhibitor of mixed lineage kinase domain-like protein (MLKL) and gasdermin D (GSDMD).
  • Interferes with executor oligomerization to inhibit necroptosis and pyroptosis.
  • Suitable for research on neurodegeneration, ischemia-reperfusion injury, and inflammatory disorders.
  • Reported molecular formula C18H15N5O6S2 and molecular weight 461.47.
  • High reported purity (98.8%) and multiple package sizes for experimental flexibility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.