Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC E/Z-necrosulfonamide | 432531-71-0 | MFCD02370191 | 98.8% | 461.47 g/mol | C18H15N5O6S2 | 5 MG
SDP

Catalog No. 5000213432
Encompass_Preferred

Racemic necrosulfonamide is a research-use small molecule that inhibits MLKL and Gasdermin D, blocking necroptosis and pyroptosis. It is supplied as a solid for biochemical and cell-based studies and should be handled according to the safety data sheet.

  • Inhibits MLKL and Gasdermin D, blocking necroptosis and pyroptosis.
  • High purity: 98.8%.
  • Molecular weight 461.47 g/mol; formula C18H15N5O6S2.
  • Appearance: light yellow to yellow solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Intended for research use only; not for human or animal consumption.

Catalog No. 50-002-13432 Supplier Medchemexpress LLC Supplier No. HY100573A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Racemic necrosulfonamide is a research-use small molecule that inhibits MLKL and Gasdermin D, blocking necroptosis and pyroptosis. It is supplied as a solid for biochemical and cell-based studies and should be handled according to the safety data sheet.

  • Inhibits MLKL and Gasdermin D, blocking necroptosis and pyroptosis.
  • High purity: 98.8%.
  • Molecular weight 461.47 g/mol; formula C18H15N5O6S2.
  • Appearance: light yellow to yellow solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Intended for research use only; not for human or animal consumption.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.