Learn More
Medchemexpress LLC E/Z-SIB-1893 | 6266-99-5 | 99.8% | 195.26 g/mol | C14H13N | 25 MG
SDP
Catalog No. 5000248451
Encompass_Preferred
(E/Z)-SIB-1893 is the racemic mixture of the (E)- and (Z)-isomers of SIB-1893, a selective, non-competitive antagonist of the metabotropic glutamate receptor subtype 5 (mGluR5). It is provided as a 25 mg research sample with reported molecular formula C14H13N, molecular weight 195.26 g/mol, and purity 99.8%.
  • Racemic mixture of E and Z isomers for receptor pharmacology studies.
  • Selective non-competitive mGluR5 antagonist useful in mGluR5-related assays.
  • High reported purity (99.8%) suitable for in vitro experiments.
  • Supplied as a 25 mg quantity for small-scale research applications.
  • Molecular formula C14H13N and molecular weight 195.26 g/mol provided for reference.
Catalog No. 50-002-48451 Supplier Medchemexpress LLC Supplier No. HY10209425MG
(E/Z)-SIB-1893 is the racemic mixture of the (E)- and (Z)-isomers of SIB-1893, a selective, non-competitive antagonist of the metabotropic glutamate receptor subtype 5 (mGluR5). It is provided as a 25 mg research sample with reported molecular formula C14H13N, molecular weight 195.26 g/mol, and purity 99.8%.
  • Racemic mixture of E and Z isomers for receptor pharmacology studies.
  • Selective non-competitive mGluR5 antagonist useful in mGluR5-related assays.
  • High reported purity (99.8%) suitable for in vitro experiments.
  • Supplied as a 25 mg quantity for small-scale research applications.
  • Molecular formula C14H13N and molecular weight 195.26 g/mol provided for reference.