Learn More
Medchemexpress LLC (E/Z)-SIB-1893 | 6266-99-5 | MFCD02262120 | 99.8% | 195.26 g/mol | C14H13N | 10 MG
SDP
Catalog No. 5000248333
Encompass_Preferred
(E/Z)-SIB-1893 is the racemic mixture of the E and Z isomers of SIB-1893, a selective, non-competitive antagonist of the metabotropic glutamate receptor subtype 5 (mGluR5) used in research to modulate glutamatergic signaling.
  • Racemic mixture of (E)- and (Z)-isomers.
  • Selective non-competitive mGluR5 antagonist.
  • Chemical formula C14H13N; molecular weight 195.26 g/mol.
  • High purity (99.8%) solid, available in mg-scale quantities and as a 10 mM solution in DMSO.
  • Store powder at -20°C for long term or 4°C for short term; store solutions at -80°C.
Catalog No. 50-002-48333 Supplier Medchemexpress LLC Supplier No. HY10209410MG
(E/Z)-SIB-1893 is the racemic mixture of the E and Z isomers of SIB-1893, a selective, non-competitive antagonist of the metabotropic glutamate receptor subtype 5 (mGluR5) used in research to modulate glutamatergic signaling.
  • Racemic mixture of (E)- and (Z)-isomers.
  • Selective non-competitive mGluR5 antagonist.
  • Chemical formula C14H13N; molecular weight 195.26 g/mol.
  • High purity (99.8%) solid, available in mg-scale quantities and as a 10 mM solution in DMSO.
  • Store powder at -20°C for long term or 4°C for short term; store solutions at -80°C.