Learn More
Medchemexpress LLC (E/Z)-SIS3 free base | 1009104-85-1 | MFCD24368989 | 99.5% | 453.53 | C28H27N3O3 | 100mg
SDP
Catalog No. 5000270452
Encompass_Preferred
E/Z-SIS3 free base exhibits an inhibitory efficacy against smad3 exhibits an anti-fibrotic and anti-inflammatory effect through a TGF- /smad3 signaling pathway[1]
Catalog No. 50-002-70452 Supplier Medchemexpress LLC Supplier No. HYW803134100MG
E/Z-SIS3 free base exhibits an inhibitory efficacy against smad3 exhibits an anti-fibrotic and anti-inflammatory effect through a TGF- /smad3 signaling pathway[1]