Learn More
Medchemexpress LLC (E/Z)-Sulindac sulfide | 32004-67-4 | 99.88% | 50 MG
SDP
Catalog No. 5000412804
Encompass_Preferred
(E/Z)-Sulindac sulfide is a potent γ-secretase modulator (GSM) that selectively reduces Aβ42 production in favor of shorter Aβ species. It can be used for researching Alzheimer's disease.
  • Potent γ-secretase modulator (GSM)
  • Selectively reduces Aβ42 production
  • Used for researching Alzheimer's disease
Catalog No. 50-004-12804 Supplier Medchemexpress LLC Supplier No. HYB1786A50MG
(E/Z)-Sulindac sulfide is a potent γ-secretase modulator (GSM) that selectively reduces Aβ42 production in favor of shorter Aβ species. It can be used for researching Alzheimer's disease.
  • Potent γ-secretase modulator (GSM)
  • Selectively reduces Aβ42 production
  • Used for researching Alzheimer's disease