Learn More
Medchemexpress LLC (E/Z)-Sulindac sulfide | 32004-67-4 | MFCD00869764 | 99.88% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB1786A
Encompass_Preferred
(E/Z)-Sulindac sulfide is a potent γ-secretase modulator (GSM) that selectively reduces Aβ42 production in favor of shorter Aβ species. It can be used for researching Alzheimer's disease.
  • Potent γ-secretase modulator (GSM).
  • Selectively reduces Aβ42 production.
  • Useful for Alzheimer's disease research.
Catalog No. 50-004-14256