Learn More
Medchemexpress LLC (E/Z)-Teriflunomide | 108605-62-5 | 99.9% | 270.21 | 10 MG
SDP
Catalog No. 5000344827
Encompass_Preferred
(E/Z)-Teriflunomide is the active metabolite of Leflunomide. Leflunomide is an immunomodulatory agent that may exert effects by inhibiting the mitochondrial enzyme dihydroorotate dehydrogenase (DHODH). It can be used for the research of rheumatoid arthritis (RA).
  • Active metabolite of Leflunomide
  • Immunomodulatory agent
  • Inhibits mitochondrial enzyme dihydroorotate dehydrogenase (DHODH)
  • Suitable for rheumatoid arthritis (RA) research
Catalog No. 50-003-44827 Supplier Medchemexpress LLC Supplier No. HY11015910MM/1ML
(E/Z)-Teriflunomide is the active metabolite of Leflunomide. Leflunomide is an immunomodulatory agent that may exert effects by inhibiting the mitochondrial enzyme dihydroorotate dehydrogenase (DHODH). It can be used for the research of rheumatoid arthritis (RA).
  • Active metabolite of Leflunomide
  • Immunomodulatory agent
  • Inhibits mitochondrial enzyme dihydroorotate dehydrogenase (DHODH)
  • Suitable for rheumatoid arthritis (RA) research