Learn More
TARGETMOL CHEMICALS INC E/Z-TG003 10MG
SDP
Catalog No. 502577280
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (E/Z)-TG003 is a potent and ATP-competitive Cdc2-like kinase (Clk) inhibitor. purity: 99%
Catalog No. 50-257-7280 Supplier TARGETMOL CHEMICALS INC Supplier No. T190110MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (E/Z)-TG003 is a potent and ATP-competitive Cdc2-like kinase (Clk) inhibitor. purity: 99%