Learn More
Medchemexpress LLC (E/Z)-Triprolidine-d8 hydrochloride | 1795134-06-3 | 1 MG
SDP
Catalog No. 5000412850
Encompass_Preferred
(E/Z)-Triprolidine-d8 hydrochloride is a deuterium-labeled version of (E/Z)-Triprolidine hydrochloride. Deuteration of drug molecules, often with stable heavy isotopes, can affect their pharmacokinetic and metabolic profiles, making them valuable as tracers for quantitation during drug development.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-12850 Supplier Medchemexpress LLC Supplier No. HYB1808S1MG
(E/Z)-Triprolidine-d8 hydrochloride is a deuterium-labeled version of (E/Z)-Triprolidine hydrochloride. Deuteration of drug molecules, often with stable heavy isotopes, can affect their pharmacokinetic and metabolic profiles, making them valuable as tracers for quantitation during drug development.
  • Used as a tracer.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.