Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E/Z)-Zinc09659342 | 591726-26-0 | 99.7% | 440.37 | C23H15F3N2O4 | 5 MG
SDP

Catalog No. 5000221395
Encompass_Preferred

(E/Z)-ZINC09659342 is a small-molecule research reagent that inhibits the Lbc-RhoA protein-protein interaction. It is supplied as a solid powder and as DMSO solutions for laboratory use; analytical documents (datasheet, COA, SDS) are available. For research use only; not for human or clinical applications.

  • Inhibits Lbc-RhoA interaction for studies of RhoA signaling.
  • High purity (approximately 99.7%).
  • Molecular weight 440.37; formula C23H15F3N2O4.
  • Available as solid quantities (5 mg-100 mg) and 10 mM DMSO solution.
  • Datasheet, certificate of analysis, and safety data sheet available.
  • Intended for laboratory research; not for clinical use.

Catalog No. 50-002-21395 Supplier Medchemexpress LLC Supplier No. HY145915A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(E/Z)-ZINC09659342 is a small-molecule research reagent that inhibits the Lbc-RhoA protein-protein interaction. It is supplied as a solid powder and as DMSO solutions for laboratory use; analytical documents (datasheet, COA, SDS) are available. For research use only; not for human or clinical applications.

  • Inhibits Lbc-RhoA interaction for studies of RhoA signaling.
  • High purity (approximately 99.7%).
  • Molecular weight 440.37; formula C23H15F3N2O4.
  • Available as solid quantities (5 mg-100 mg) and 10 mM DMSO solution.
  • Datasheet, certificate of analysis, and safety data sheet available.
  • Intended for laboratory research; not for clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.